Assay Details
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199410.2 | NP_955442.1 | 6 - 7 | 736 | 86 | |
GenBank mRNA | BC063180.1 | - | 6 - 7 | 740 | 6920424 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
