Assay Details
Gene Symbol: B3galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105887.1 | NP_001099357.1 | 3 | 5206 | 128 | |
XM_008768569.1 | XP_008766791.1 | 4 | 7220 | 128 | ||
XM_008768570.1 | XP_008766792.1 | 4 | 5317 | 128 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132199 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127005 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132199 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132194 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132194 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132199 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132199 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132194 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P178498 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8726 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32111 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10750490 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10750489 | - | - |
Rat Genome 230 2.0 Array | 1375030_at | - | - |
Rat Genome 230 2.0 Array | 1375030_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375030_at | - | - |
Affymetrix HT Rat Focus Array | 1375030_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100001740.rn.1 | TC1100000355.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001745.rn.1 | TC1100000355.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001743.rn.1 | TC1100000355.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001737.rn.1 | TC1100000355.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001741.rn.1 | TC1100000355.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001744.rn.1 | TC1100000355.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001736.rn.1 | TC1100000355.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001742.rn.1 | TC1100000355.rn.1 | RTA TC |
Rat Expression Set 230 | 1375030_at | - | - |
Rat Expression Set 230 | 1375030_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750489 | - | - |
Rat Genome U34 Set | rc_AI639285_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20860779 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363983 | - | - |
