Assay Details
Gene Symbol: Mc2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100491.1 | NP_001093961.1 | 2 | 871 | 126 | |
XM_006254893.2 | XP_006254955.1 | 2 | 919 | 126 | ||
XM_008772091.1 | XP_008770313.1 | 3 | 970 | 126 | ||
XM_008772092.1 | XP_008770314.1 | 3 | 966 | 126 | ||
XM_008772093.1 | XP_008770315.1 | 3 | 1964 | 126 | ||
XM_008772094.1 | XP_008770316.1 | 2 | 1913 | 126 | ||
XM_008772095.1 | XP_008770317.1 | 3 | 1917 | 126 | ||
GenBank mRNA | AF547168.1 | - | 1 | 529 | 126 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
