Assay Details
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107142.1 | NP_001100612.1 | 12 | 2342 | 145 | |
XM_006249365.1 | XP_006249427.1 | 11 | 2260 | 145 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P142318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162713 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P142318 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P142318 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P142318 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P142318 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92395 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10762015 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762020 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762014 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762019 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762018 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762016 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762017 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762010 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10762009 | - | - |
Rat Genome 230 2.0 Array | 1375998_at | - | - |
Rat Genome 230 2.0 Array | 1375998_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375998_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375998_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200013548.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013552.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013551.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013541.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013550.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013553.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013540.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013539.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013542.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013544.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013545.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013547.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013543.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013546.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013549.rn.1 | TC1200001592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200013556.rn.1 | TC1200001592.rn.1 | RTA TC |
Rat Expression Set 230 | 1375998_at | - | - |
Rat Expression Set 230 | 1375998_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10762009 | - | - |
Rat Genome U34 Set | rc_AI059152_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21239972 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354823 | - | - |
