Assay Details
| Gene Symbol: Arid5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244806.2 | XP_006244868.1 | 7 | 4516 | 157 | |
| XM_006244807.2 | XP_006244869.1 | 7 | 4513 | 157 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arid5a
Genomic Map