Assay Details
| Gene Symbol: B3gnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107240.1 | NP_001100710.1 | 3 | 2765 | 141 | |
| XM_006251554.2 | XP_006251616.1 | 2 | 2564 | 141 | ||
| XM_006251555.1 | XP_006251617.1 | 2 | 2448 | 141 | ||
| XM_006251556.2 | XP_006251618.1 | 2 | 2447 | 141 | ||
| XM_008770427.1 | XP_008768649.1 | 2 | 4283 | 141 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B3gnt2
Genomic Map