Assay Details
Gene Symbol: Zmym5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728259.4 | XP_002728305.2 | 8 | 2624 | 150 | |
XM_006252082.2 | XP_006252144.1 | 7 | 2528 | 150 | ||
XM_006252083.2 | XP_006252145.1 | 7 | 2464 | 150 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
