Assay Details
| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107239.1 | NP_001100709.1 | 20 | 3137 | 131 | |
| XM_008770320.1 | XP_008768542.1 | 22 | 3367 | 131 | ||
| XM_008770321.1 | XP_008768543.1 | 21 | 3295 | 131 | ||
| XM_008770322.1 | XP_008768544.1 | 20 | 2716 | 131 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
cAMP biosynthetic process
axonogenesis
long-term memory
circadian rhythm
response to lithium ion
intracellular signal transduction
response to drug
regulation of circadian rhythm
nucleotide binding
adenylate cyclase activity
calcium- and calmodulin-responsive adenylate cyclase activity
cAMP biosynthetic process
axonogenesis
long-term memory
circadian rhythm
response to lithium ion
intracellular signal transduction
response to drug
regulation of circadian rhythm
nucleotide binding
adenylate cyclase activity
calcium- and calmodulin-responsive adenylate cyclase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P232585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118034 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P232585 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P232585 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P232585 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P232585 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44371 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38212 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10774133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774123 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774121 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774140 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774122 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774137 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10774132 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10774120 | - | - |
| Rat Genome 230 2.0 Array | 1378566_at | - | - |
| Rat Genome 230 2.0 Array | 1378566_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400007169.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007156.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007160.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007167.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007168.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007157.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007161.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007154.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007165.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007150.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007158.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007164.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007148.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007153.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007170.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007147.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007152.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007155.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007149.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007151.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007163.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400007166.rn.1 | TC1400000969.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10774120 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21112309 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366411 | - | - |

Adcy1
Genomic Map