Assay Details
Gene Symbol: Abhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106275.1 | NP_001099745.1 | 11 | 1969 | 112 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P041421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139368 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041421 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041421 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P041421 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10329 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P900209 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37017 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16547 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3458 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46696 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10708059 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708061 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708055 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708063 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708057 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708064 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708056 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708054 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708058 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708062 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708060 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10708053 | - | - |
Rat Genome 230 2.0 Array | 1390455_at | - | - |
Rat Genome 230 2.0 Array | 1390455_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398109_at | - | - |
Rat Genome 230 2.0 Array | 1398109_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390455_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390455_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100014322.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014329.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014332.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014336.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014324.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014327.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014330.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014331.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014323.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014325.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014326.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014328.rn.1 | TC0100001826.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100014335.rn.1 | TC0100001826.rn.1 | RTA TC |
Rat Expression Set 230 | 1390455_at | - | - |
Rat Expression Set 230 | 1390455_PM_at | - | - |
Rat Expression Set 230 | 1398109_at | - | - |
Rat Expression Set 230 | 1398109_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708053 | - | - |
Rat Genome U34 Set | rc_AI013474_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22104079 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354232 | - | - |
