Assay Details
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013973.1 | NP_001013995.1 | 7 | 118 | ||
XM_006247579.2 | XP_006247641.1 | 7 | 118 | |||
XM_006247580.2 | XP_006247642.1 | 8 | 118 | |||
GenBank mRNA | BC083567.1 | - | 7 | 7112851 | 118 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
