Assay Details
| Gene Symbol: LOC102555919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246356.2 | XP_006246418.1 | 5 | 4355 | 92 | |
| XM_008767728.1 | XP_008765950.1 | 4 | 2318 | 92 | ||
| XM_008767729.1 | XP_008765951.1 | 5 | 4540 | 92 | ||
| XM_008767730.1 | XP_008765952.1 | 4 | 2315 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102555919
Genomic Map