Assay Details
| Gene Symbol: Neurod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019326.1 | NP_062199.1 | 1 | 1642 | 133 | |
| GenBank mRNA | D82868.1 | - | 1 | 1642 | 133 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Neurod2
Genomic Map