Assay Details
| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007666.1 | NP_001007667.1 | 5 - 6 | 1023 | 158 | |
| GenBank mRNA | BC083660.1 | - | 5 - 6 | 1023 | 7135331 | 158 |
More Information
Updated Mapping Information
Set Membership: |
|

Bcs1l
Genomic Map