Assay Details
| Gene Symbol: Krt75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008828.1 | NP_001008828.1 | 3 - 4 | 752 | 162 | |
| XM_006242325.2 | XP_006242387.1 | 3 - 4 | 952 | 162 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt75
Genomic Map