Assay Details
Gene Symbol: Bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009524.1 | NP_001009524.1 | 20 - 21 | 2165 | 125 | |
XM_006242737.2 | XP_006242799.1 | 20 - 21 | 2043 | 125 | ||
GenBank mRNA | AF299297.1 | - | 20 - 21 | 2165 | 125 | |
AF299298.1 | - | 20 - 21 | 2019 | 125 | ||
AF414431.1 | - | 20 - 21 | 2019 | 125 | ||
AY040870.1 | - | 20 - 21 | 2165 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
