Assay Details
Gene Symbol: Bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009524.1 | NP_001009524.1 | 7 - 8 | 916 | 129 | |
XM_006242737.2 | XP_006242799.1 | 7 - 8 | 794 | 129 | ||
GenBank mRNA | AF299297.1 | - | 7 - 8 | 916 | 129 | |
AF299298.1 | - | 7 - 8 | 770 | 129 | ||
AF414431.1 | - | 7 - 8 | 770 | 129 | ||
AY040870.1 | - | 7 - 8 | 916 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
