Assay Details
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009623.1 | NP_001009623.1 | 4 - 5 | 745 | 131 | |
XM_006246625.2 | XP_006246687.1 | 5 - 6 | 1330 | 131 | ||
XM_008767796.1 | XP_008766018.1 | 4 - 5 | 1486 | 131 | ||
XM_008767797.1 | XP_008766019.1 | 4 - 5 | 1483 | 131 | ||
XM_008767798.1 | XP_008766020.1 | 3 - 4 | 1438 | 131 | ||
XM_008767799.1 | XP_008766021.1 | 3 - 4 | 1435 | 131 | ||
XM_008767800.1 | XP_008766022.1 | 4 - 5 | 1486 | 131 | ||
XM_008767801.1 | XP_008766023.1 | 4 - 5 | 1483 | 131 | ||
XM_008767802.1 | XP_008766024.1 | 3 - 4 | 1438 | 131 | ||
GenBank mRNA | BC087135.1 | - | 4 - 5 | 745 | 7123934 | 131 |
FQ215356.1 | - | 4 - 5 | 515 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
