Assay Details
| Gene Symbol: Zfp414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009664.1 | NP_001009664.1 | 4 - 5 | 610 | 101 | |
| XM_006241117.2 | XP_006241179.1 | 4 - 5 | 800 | 101 | ||
| XM_006241119.2 | XP_006241181.1 | 4 - 5 | 763 | 101 | ||
| XM_006241121.2 | XP_006241183.1 | 4 - 5 | 798 | 101 | ||
| GenBank mRNA | BC087693.1 | - | 4 - 5 | 610 | 7310348 | 101 |
| FQ220382.1 | - | 4 - 5 | 654 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp414
Genomic Map