Assay Details
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009674.1 | NP_001009674.1 | 2 | 316 | 132 | |
GenBank mRNA | BC087127.1 | - | 2 | 316 | 7111761 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
