Assay Details
| Gene Symbol: Ugt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031533.5 | NP_113721.4 | 4 - 5 | 1126 | 150 | |
| GenBank mRNA | BC089792.1 | - | 4 - 5 | 1109 | 7370057 | 150 |
| DQ351215.1 | - | 4 - 5 | 1467 | 150 | ||
| X03478.1 | - | 4 - 5 | 1015 | 150 | ||
| Gene Symbol: Ugt2b17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769997.1 | XP_008768219.1 | 4 - 5 | 1466 | 150 | |
| XM_008769998.1 | XP_008768220.1 | 4 - 5 | 2223 | 150 | ||
| XM_008769999.1 | XP_008768221.1 | 4 - 5 | 1120 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ugt2b
Genomic Map