Assay Details
| Gene Symbol: Mug2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002826.1 | NP_001002826.1 | 3893 | 123 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P536097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73895 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P536097 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P536097 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P536097 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P153947 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P536097 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53468 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89641 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99525 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57570 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10858479 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858474 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858481 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858472 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858484 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858477 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858471 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858486 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858475 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858478 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858482 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858485 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858480 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858473 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10858470 | - | - |
| Rat Genome 230 2.0 Array | 1370027_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370027_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1388229_a_at | - | - |
| Rat Genome 230 2.0 Array | 1388229_PM_a_at | - | - |
| Rat Expression Set 230 | 1370027_a_at | - | - |
| Rat Expression Set 230 | 1370027_PM_a_at | - | - |
| Rat Expression Set 230 | 1388229_PM_a_at | - | - |
| Rat Expression Set 230 | 1388229_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858470 | - | - |
| Rat Genome U34 Set | M22993cds_s_at | - | - |
| Rat Genome U34 Set | M22360_s_at | - | - |
| Rat Genome U34 Set | M22359mRNA_s_at | - | - |
| Rat Genome U34 Set | J03524_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374541 | - | - |

Mug2
Genomic Map