Assay Details
| Gene Symbol: tGap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007635.2 | NP_001007636.2 | 19 - 20 | 2685 | 137 | |
| XM_006232134.2 | XP_006232196.1 | 21 - 22 | 3027 | 137 | ||
| GenBank mRNA | AY631396.1 | - | 19 - 20 | 2685 | 137 | |
| AY631398.1 | - | 12 - 13 | 1829 | 137 | ||
| AY631399.1 | - | 19 - 20 | 2450 | 137 | ||
| AY631400.1 | - | 15 - 16 | 1824 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

tGap1
Genomic Map