Assay Details
| Gene Symbol: tGap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007635.2 | NP_001007636.2 | 22 - 23 | 3243 | 125 | |
| XM_006232134.2 | XP_006232196.1 | 24 - 25 | 3585 | 125 | ||
| GenBank mRNA | AY631396.1 | - | 22 - 23 | 3243 | 125 | |
| AY631398.1 | - | 15 - 16 | 2387 | 125 | ||
| AY631399.1 | - | 22 - 23 | 3008 | 125 | ||
| AY631400.1 | - | 18 - 19 | 2382 | 125 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

tGap1
Genomic Map