Assay Details
| Gene Symbol: Zbtb38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012471.1 | NP_001012489.1 | 1 | 3574 | 105 | |
| XM_006243578.2 | XP_006243640.1 | 2 | 6350 | 105 | ||
| XM_006243579.2 | XP_006243641.1 | 3 | 4089 | 105 | ||
| XM_006243580.2 | XP_006243642.1 | 5 | 4215 | 105 | ||
| XM_006243582.2 | XP_006243644.1 | 3 | 4018 | 105 | ||
| XM_006243584.1 | XP_006243646.1 | 3 | 3672 | 105 | ||
| XM_006243589.2 | XP_006243651.1 | 1 | 3513 | 105 | ||
| XM_008766520.1 | XP_008764742.1 | 3 | 3900 | 105 | ||
| XM_008766521.1 | XP_008764743.1 | 6 | 4374 | 105 | ||
| XM_008766522.1 | XP_008764744.1 | 7 | 4385 | 105 | ||
| XM_008766523.1 | XP_008764745.1 | 5 | 4270 | 105 | ||
| XM_008766524.1 | XP_008764746.1 | 4 | 4149 | 105 | ||
| GenBank mRNA | AY623002.1 | - | 1 | 3574 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zbtb38
Genomic Map