Assay Details
| Gene Symbol: Klri2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012648.1 | NP_001012666.1 | 3 - 4 | 406 | 70 | |
| GenBank mRNA | AY324871.1 | - | 3 - 4 | 406 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klri2
Genomic Map