Assay Details
| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017487.1 | NP_001017487.1 | 1 - 2 | 249 | 85 | |
| GenBank mRNA | BC085334.1 | - | 1 - 2 | 249 | 7301029 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Bri3bp
Genomic Map