Assay Details
Gene Symbol: Hyal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024321.1 | NP_001019492.1 | 3 - 4 | 1351 | 127 | |
XM_006236166.2 | XP_006236228.1 | 4 - 5 | 1469 | 127 | ||
XM_008762806.1 | XP_008761028.1 | 5 - 6 | 1697 | 127 | ||
XM_008762807.1 | XP_008761029.1 | 4 - 5 | 1604 | 127 | ||
XM_008762808.1 | XP_008761030.1 | 4 - 5 | 1519 | 127 | ||
GenBank mRNA | BC079447.1 | - | 5 - 6 | 1674 | 127 | |
BC091219.1 | - | 3 - 4 | 1351 | 7112082 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
