Assay Details
Gene Symbol: Bles03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024233.2 | NP_001019404.2 | 2 | 880 | 76 | |
NM_001243534.1 | NP_001230463.1 | 2 | 894 | 76 | ||
GenBank mRNA | AF502571.2 | - | 1 | 87 | 76 | |
BC093387.1 | - | 2 | 739 | 7389419 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
