Assay Details
Gene Symbol: Cmtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029914.1 | NP_001025085.1 | 2 - 3 | 995 | 114 | |
GenBank mRNA | BC098029.1 | - | 2 - 3 | 995 | 7456234 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
