Assay Details
| Gene Symbol: C4bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012516.2 | NP_036648.2 | 8 - 9 | 1158 | 104 | |
| GenBank mRNA | BC088165.1 | - | 8 - 9 | 1158 | 7324823 | 104 |
| Z50051.1 | - | 6 - 7 | 821 | 104 | ||
More Information
Set Membership: |
|

C4bpa
Genomic Map