Assay Details
Gene Symbol: Ren | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012642.4 | NP_036774.4 | 4 - 5 | 507 | 65 | |
GenBank mRNA | BC078878.1 | - | 4 - 5 | 507 | 7126969 | 65 |
J02941.1 | - | 4 - 5 | 700 | 65 | ||
S60054.1 | - | 4 - 5 | 517 | 65 | ||
X07033.1 | - | 4 - 5 | 517 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
