Assay Details
| Gene Symbol: Gpr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037359.1 | NP_001032436.1 | 1 | 876 | 79 | |
| XM_006245537.2 | XP_006245599.1 | 2 | 1019 | 79 | ||
| XM_006245539.2 | XP_006245601.1 | 2 | 1234 | 79 | ||
| XM_006245540.2 | XP_006245602.1 | 2 | 1231 | 79 | ||
| GenBank mRNA | AB240684.1 | - | 1 | 876 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr35
Genomic Map