Assay Details
| Gene Symbol: Bles03 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243534.1 | NP_001230463.1 | 1 - 2 | 231 | 77 | |
| GenBank mRNA | BC093387.1 | - | 1 - 2 | 76 | 7389419 | 77 |
| CK483724.1 | - | 1 - 2 | 73 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Dr1 associated protein 1 (negative cofactor 2 alpha) | Drap1 | -87.0 | Anti-sense | Chr. 1 - 220803783 - 220806476 on NCBI Build Rnor_6.0 | |
| Downstream | fos-like antigen 1 | Fosl1 |
Fra-1, fra-1 |
17.7kb | Sense | Chr. 1 - 220826560 - 220835066 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC093387.1 transcripts.
Set Membership: |
|

Bles03
Genomic Map