Assay Details
Gene Symbol: Chrm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017362.4 | NP_059058.1 | 2229 | 92 | ||
GenBank mRNA | M22926.1 | - | 2229 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P036484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151731 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036484 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036484 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P036484 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11937 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11832 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11803 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43980 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37877 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79885 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10848167 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848169 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10848165 | - | - |
Rat Genome 230 2.0 Array | 1369786_at | - | - |
Rat Genome 230 2.0 Array | 1369786_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369786_at | - | - |
Affymetrix HT Rat Focus Array | 1369786_PM_at | - | - |
Rat Expression Set 230 | 1369786_at | - | - |
Rat Expression Set 230 | 1369786_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848165 | - | - |
Rat Genome U34 Set | M22926mRNA_at | - | - |
Rat Genome U34 Set | M22926mRNA_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21566173 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355921 | - | - |
RatRef-12 Expression BeadChip | ILMN_60205 | - | - |
