Assay Details
Gene Symbol: Kcna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012970.3 | NP_037102.1 | 3 | 1909 | 123 | |
XM_006233132.2 | XP_006233194.1 | 3 | 2495 | 123 | ||
XM_006233133.2 | XP_006233195.1 | 3 | 2077 | 123 | ||
XM_006233134.2 | XP_006233196.1 | 3 | 2060 | 123 | ||
XM_006233135.2 | XP_006233197.1 | 3 | 1864 | 123 | ||
XM_008761371.1 | XP_008759593.1 | 3 | 2105 | 123 | ||
GenBank mRNA | J04731.1 | - | 3 | 1909 | 123 | |
M74449.1 | - | 1 | 1378 | 123 | ||
X16003.1 | - | 3 | 1890 | 123 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
