Assay Details
| Gene Symbol: Gstp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012577.2 | NP_036709.1 | 5 | 336 | 72 | |
| GenBank mRNA | BC058439.1 | - | 5 | 340 | 6889675 | 72 |
| BC058440.1 | - | 5 | 340 | 6917961 | 72 | |
| FJ179407.1 | - | 4 | 266 | 72 | ||
| FQ217279.1 | - | 5 | 336 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gstp1
Genomic Map