Assay Details
| Gene Symbol: Glrx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013034.1 | NP_001013052.1 | 4 - 5 | 475 | 118 | |
| XM_006249955.1 | XP_006250017.1 | 3 - 4 | 514 | 118 | ||
| XM_006249956.2 | XP_006250018.1 | 3 - 4 | 537 | 118 | ||
| XM_006249957.2 | XP_006250019.1 | 3 - 4 | 336 | 118 | ||
| XM_006249958.2 | XP_006250020.1 | 3 - 4 | 404 | 118 | ||
| XM_008769523.1 | XP_008767745.1 | 4 - 5 | 472 | 118 | ||
| XM_008769524.1 | XP_008767746.1 | 3 - 4 | 364 | 118 | ||
| XM_008769525.1 | XP_008767747.1 | 3 - 4 | 420 | 118 | ||
| XM_008769526.1 | XP_008767748.1 | 3 - 4 | 404 | 118 | ||
| GenBank mRNA | BC079292.1 | - | 4 - 5 | 475 | 7134405 | 118 |
| FQ213555.1 | - | 3 - 4 | 280 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glrx2
Genomic Map