Assay Details
Gene Symbol: LOC498414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY539948.1 | - | 774 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
