Assay Details
| Gene Symbol: LOC499541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047942.1 | NP_001041407.1 | 1 - 2 | 83 | 129 | |
| XM_008760791.1 | XP_008759013.1 | 4 - 5 | 1011 | 129 | ||
| GenBank mRNA | AY383700.1 | - | 1 - 2 | 83 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC499541
Genomic Map