Assay Details
| Gene Symbol: LOC499584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047948.1 | NP_001041413.1 | 1 - 2 | 37 | 70 | |
| GenBank mRNA | AY539953.1 | - | 1 - 2 | 37 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC499584
Genomic Map