Assay Details
| Gene Symbol: RGD1564345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077681.1 | NP_001071149.1 | 2 | 762 | 131 | |
| GenBank mRNA | BC127528.1 | - | 2 | 762 | 7114751 | 131 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1564345
Genomic Map