Assay Details
| Gene Symbol: RGD1562638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100944.1 | NP_001094414.1 | 2 - 3 | 2355 | 76 | |
| XM_008771316.1 | XP_008769538.1 | 6 - 7 | 3161 | 76 | ||
| XM_008771317.1 | XP_008769539.1 | 5 - 6 | 3109 | 76 | ||
| XM_008771318.1 | XP_008769540.1 | 7 - 8 | 3215 | 76 | ||
| XM_008771319.1 | XP_008769541.1 | 6 - 7 | 3161 | 76 | ||
| XM_008771320.1 | XP_008769542.1 | 4 - 5 | 3375 | 76 | ||
| XM_008771321.1 | XP_008769543.1 | 3 - 4 | 2788 | 76 | ||
| XM_008771322.1 | XP_008769544.1 | 6 - 7 | 3161 | 76 | ||
| GenBank mRNA | BC128761.1 | - | 2 - 3 | 2355 | 7113007 | 76 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1562638
Genomic Map