Assay Details
| Gene Symbol: MT-ND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | NC_001665.ND6.0 | - | 446 | 77 | ||
More Information
Additional Information:
NC_001665 (strand:-; 13543..14061); mitochondrially encoded NADH dehydrogenase 6
Set Membership: |
|

MT-ND6
Genomic Map