Assay Details
| Gene Symbol: Cited4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053699.1 | NP_446151.1 | 1 | 43 | 110 | |
| GenBank mRNA | AF361477.1 | - | 1 | 43 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AF361477.1,NM_053699.1 transcripts.
This assay is in the 5'UTR region of the AF361477.1,NM_053699.1 transcripts.
Set Membership: |
|

Cited4
Genomic Map