Assay Details
| Gene Symbol: Fam181b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109150.2 | NP_001102620.1 | 1 | 1245 | 99 | |
| GenBank mRNA | BC158703.1 | - | 1 | 1245 | 9029854 | 99 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam181b
Genomic Map