Assay Details
Gene Symbol: Vom2r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099487.1 | NP_001092957.1 | 5 | 1850 | 144 |
Gene Symbol: Vom2r38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099476.1 | NP_001092946.1 | 2069 | 144 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P048334 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P125362 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P094696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151888 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125362 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P048334 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P094696 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125362 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P048334 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P094696 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P048334 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P125362 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P094696 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P438327 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31342 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102663 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10706287 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706283 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706279 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706286 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706282 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706280 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706278 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706288 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706277 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706285 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706284 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10706276 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10706276 | - | - |
Affymetrix Rat Clariom S Assay | PSR0100042897.rn.1 | TC0100005320.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100042898.rn.1 | TC0100005320.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100042888.rn.1 | TC0100005320.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10706276 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10706276 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21753547 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348833 | - | - |
