Assay Details
| Gene Symbol: Mirlet7i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031808.1 | 1 | 45 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MON2 homolog (S. cerevisiae) | Mon2 | RGD1308808 | -8.7kb | Sense | Chr. 7 - 66717470 - 66794041 on NCBI Build Rnor_6.0 |
| Downstream |
protein phosphatase, Mg2+/Mn2+ dependent, 1H |
Ppm1h | 42.9kb | Anti-sense | Chr. 7 - 66845692 - 67117317 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7i
Genomic Map