Assay Details
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 30B | Tmem30b | -20.8kb | Sense | Chr. 6 - 96439607 - 96442682 on NCBI Build Rnor_6.0 | |
Downstream |
protein kinase C, eta |
Prkch | 14.7kb | Anti-sense | Chr. 6 - 96479243 - 96677368 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
