Assay Details
| Gene Symbol: Tbc1d25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106955.2 | NP_001100425.1 | 3 - 4 | 439 | 100 | |
| XM_006256701.2 | XP_006256763.1 | 3 - 4 | 533 | 100 | ||
| GenBank mRNA | BC161862.1 | - | 3 - 4 | 439 | 7097051 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d25
Genomic Map