Assay Details
| Gene Symbol: Eef1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108799.2 | NP_001102269.1 | 2 - 3 | 505 | 91 | |
| GenBank mRNA | BC168738.1 | - | 2 - 3 | 505 | 6918011 | 91 |
| FQ211150.1 | - | 2 - 3 | 274 | 91 | ||
| FQ211594.1 | - | 2 - 3 | 301 | 91 | ||
| FQ216958.1 | - | 2 - 3 | 273 | 91 | ||
| FQ217523.1 | - | 2 - 3 | 273 | 91 | ||
| FQ218320.1 | - | 2 - 3 | 273 | 91 | ||
| FQ221159.1 | - | 2 - 3 | 273 | 91 | ||
| FQ221342.1 | - | 2 - 3 | 275 | 91 | ||
| FQ221482.1 | - | 2 - 3 | 273 | 91 | ||
| FQ222690.1 | - | 2 - 3 | 274 | 91 | ||
| FQ223187.1 | - | 2 - 3 | 273 | 91 | ||
| FQ228255.1 | - | 2 - 3 | 274 | 91 | ||
| FQ228375.1 | - | 2 - 3 | 275 | 91 | ||
| FQ228595.1 | - | 2 - 3 | 274 | 91 | ||
| FQ232532.1 | - | 2 - 3 | 276 | 91 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eef1b2
Genomic Map